accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3CH3CH3OH2C

CAS RN 926-02-3

IUPAC Name: 2-(ethenyloxy)-2-methylpropane
Synonyms: 2-(ethenyloxy)-2-methylpropane
Molecular Weight (g/mol): 100.16
Molecular Formula: C6H12O
InChi Key: PGYJSURPYAAOMM-UHFFFAOYSA-N
SMILES: CC(C)(C)OC=C
Boiling Point 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)