CAS RN 924296-18-4
CAS RN 924296-18-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
DUB-IN-1, MedChemExpress
MedChemExpress DUB-IN-1 is an active inhibitor of ubiquitin-specific proteases (USPs), with an IC50 of 0.85 μM for USP8.