accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 921938-80-9

IUPAC Name: 4-methyl-2H,3H,4H-pyrido[3,2-b][1,4]oxazine-7-carbaldehyde
Synonyms: 4-methyl-2H,3H-pyrido[3,2-b][1,4]oxazine-7-carbaldehyde
Molecular Weight (g/mol): 178.19
Molecular Formula: C9H10N2O2
InChi Key: CVHLHSUUCRUZHB-UHFFFAOYSA-N
SMILES: CN1CCOC2=C1N=CC(C=O)=C2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results