accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 920966-19-4

CH3H3CCH3OOHNN

CAS RN 920966-19-4

IUPAC Name: tert-butyl N-(3-cyanocyclohexyl)carbamate
Synonyms: tert-butyl N-(3-cyanocyclohexyl)carbamate
Molecular Weight (g/mol): 224.3
Molecular Formula: C12H20N2O2
InChi Key: KODINHTWTSHDJW-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)NC1CCCC(C1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results