accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 92-31-9

CAS RN 92-31-9

IUPAC Name: 3-amino-7-(dimethylamino)-2-methyl-5λ⁴-phenothiazin-5-ylium chloride
Synonyms: toluidine blue tolonium chloride
Molecular Weight (g/mol): 305.82
Molecular Formula: C15H16ClN3S
InChi Key: HNONEKILPDHFOL-UHFFFAOYSA-M
SMILES: [Cl-].CN(C)C1=CC=C2N=C3C=C(C)C(N)=CC3=[S+]C2=C1
Molecular Weight (g/mol) 
  • (9)
  • (3)
Quantity 
  • (1)
  • (1)
  • (5)
  • (1)
  • (3)
  • (1)
Color 
  • (1)
  • (5)
Grade 
  • (3)
Percent Purity 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (3)
  • (9)
  • (3)
  • (1)
  • (1)
  • (5)
  • (1)
  • (3)
  • (1)
  • (1)
  • (5)
  • (3)
  • (3)