CAS RN 91940-87-3
CAS RN 91940-87-3
IUPAC Name:
hydrogen 3-(3-{[2-(3,4-dimethoxyphenyl)ethyl](methyl)amino}propyl)-7,8-dimethoxy-2,3,4,5-tetrahydro-1H-3-benzazepin-2-one chloride
Synonyms:
hydrogen 3-(3-{[2-(3,4-dimethoxyphenyl)ethyl](methyl)amino}propyl)-7,8-dimethoxy-4,5-dihydro-1H-3-benzazepin-2-one chloride
Molecular Weight (g/mol):
493.04
Molecular Formula:
C26H37ClN2O5
InChi Key:
ZRNKXJHEQKMWCH-UHFFFAOYSA-N
SMILES:
[H+].[Cl-].COC1=CC=C(CCN(C)CCCN2CCC3=CC(OC)=C(OC)C=C3CC2=O)C=C1OC
1
–
2
of
2
results
1
Tocris Bioscience™ Zatebradine hydrochloride
CAS: 91940-87-3 Molecular Formula: C26H37ClN2O5
2
Zatebradine hydrochloride, MedChemExpress
MedChemExpress Zatebradine (UL-FS-49 (free base)) is a potent inhibitor of hyperpolarization-activated cyclic nucleotide-gated (HCN) channels with an IC50 values 1.96 μM. Zatebradine blocks the slow inward current through human HCN1, HCN2, HCN3 and HCN4 channels, with IC50 values of 1.83 μM, 2.21 μM, 1.90 μM and 1.88 μM, respectively.