accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3SSPOOH3COH3C

CAS RN 919-86-8

IUPAC Name: dimethyl {[2-(ethylsulfanyl)ethyl]sulfanyl}phosphonate
Synonyms: demeton-S-methyl demetox metasystox J DSM metaisosystox thiometon oxon methyl demeton thioester duratox metasystox 55 methylmercaptofostiol (ussr)
Molecular Weight (g/mol): 230.28
Molecular Formula: C6H15O3PS2
InChi Key: WEBQKRLKWNIYKK-UHFFFAOYSA-N
SMILES: CCSCCSP(=O)(OC)OC
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results