accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 919-31-3

CH3OSiNOH3COH3C

CAS RN 919-31-3

IUPAC Name: 3-(triethoxysilyl)propanenitrile
Synonyms: 3-(triethoxysilyl)propanenitrile
Molecular Weight (g/mol): 217.34
Molecular Formula: C9H19NO3Si
InChi Key: GBQYMXVQHATSCC-UHFFFAOYSA-N
SMILES: CCO[Si](CCC#N)(OCC)OCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results