accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 91872-10-5

CH3NHNH2BrBr

CAS RN 91872-10-5

IUPAC Name: 2-amino-3,5-dibromo-6-methylpyridin-1-ium
Synonyms: 2-amino-3,5-dibromo-6-methylpyridin-1-ium
Molecular Weight (g/mol): 266.94
Molecular Formula: C6H7Br2N2
InChi Key: BLUYMDMBXIMIGK-UHFFFAOYSA-O
SMILES: CC1=[NH+]C(N)=C(Br)C=C1Br

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results