accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3CNCH3SCH3OO

CAS RN 918-05-8

IUPAC Name: N,N-dimethylmethanesulfonamide
Synonyms: N,N-dimethylmethanesulfonamide
Molecular Weight (g/mol): 123.17
Molecular Formula: C3H9NO2S
InChi Key: WCFDSGHAIGTEKL-UHFFFAOYSA-N
SMILES: CN(C)S(C)(=O)=O
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results