accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 916792-16-0

ONO

CAS RN 916792-16-0

IUPAC Name: 6-(cyclohexyloxy)pyridine-3-carbaldehyde
Synonyms: 6-(cyclohexyloxy)pyridine-3-carbaldehyde
Molecular Weight (g/mol): 205.26
Molecular Formula: C12H15NO2
InChi Key: KTDSNAKFASLMHF-UHFFFAOYSA-N
SMILES: O=CC1=CN=C(OC2CCCCC2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results