accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 916766-83-1

CH3NNCl

CAS RN 916766-83-1

IUPAC Name: 3-[4-(chloromethyl)phenyl]-1-methyl-1H-pyrazole
Synonyms: 3-[4-(chloromethyl)phenyl]-1-methylpyrazole
Molecular Weight (g/mol): 206.67
Molecular Formula: C11H11ClN2
InChi Key: YLJMPTWMRVXTQQ-UHFFFAOYSA-N
SMILES: CN1C=CC(=N1)C1=CC=C(CCl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results