accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 916497-18-2

NNSHO

CAS RN 916497-18-2

IUPAC Name: 2-[2-(piperidin-1-yl)-1,3-thiazol-4-yl]phenol
Synonyms: 2-[2-(piperidin-1-yl)-1,3-thiazol-4-yl]phenol
Molecular Weight (g/mol): 260.36
Molecular Formula: C14H16N2OS
InChi Key: CIZFLLFRDXZCLM-UHFFFAOYSA-N
SMILES: OC1=CC=CC=C1C1=CSC(=N1)N1CCCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results