accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 916420-47-8

FFFOHOClFFF

CAS RN 916420-47-8

IUPAC Name: 2-chloro-4,6-bis(trifluoromethyl)benzoic acid
Synonyms: 2-chloro-4,6-bis(trifluoromethyl)benzoic acid
Molecular Weight (g/mol): 292.56
Molecular Formula: C9H3ClF6O2
InChi Key: MXHOYUZOOIMVGF-UHFFFAOYSA-N
SMILES: OC(=O)C1=C(C=C(C=C1Cl)C(F)(F)F)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results