accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 916162-54-4

BOOCH3CH3H3CCH3NOCH3

CAS RN 916162-54-4

IUPAC Name: 8-methoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)quinoline
Synonyms: 8-methoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)quinoline
Molecular Weight (g/mol): 285.15
Molecular Formula: C16H20BNO3
InChi Key: UJPLJBDGXPKCDH-UHFFFAOYSA-N
SMILES: COC1=CC=C(B2OC(C)(C)C(C)(C)O2)C2=CC=CN=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results