accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3NClNH3CCl

CAS RN 91591-63-8

IUPAC Name: 2,5-dichloro-4,6-dimethylpyridine-3-carbonitrile
Synonyms: 2,5-dichloro-4,6-dimethylpyridine-3-carbonitrile
Molecular Weight (g/mol): 201.05
Molecular Formula: C8H6Cl2N2
InChi Key: UCGWYTUBYASPFG-UHFFFAOYSA-N
SMILES: CC1=NC(Cl)=C(C#N)C(C)=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results