accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3NNOOFFFFF

CAS RN 915707-42-5

IUPAC Name: 2,3,4,5,6-pentafluorophenyl 4-(1-methyl-1H-pyrazol-3-yl)benzoate
Synonyms: 2,3,4,5,6-pentafluorophenyl 4-(1-methylpyrazol-3-yl)benzoate
Molecular Weight (g/mol): 368.26
Molecular Formula: C17H9F5N2O2
InChi Key: LGHZGIHYFYLEAC-UHFFFAOYSA-N
SMILES: CN1C=CC(=N1)C1=CC=C(C=C1)C(=O)OC1=C(F)C(F)=C(F)C(F)=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results