accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 912760-91-9

IUPAC Name: ethyl 3-(2-methoxypyridin-3-yl)prop-2-enoate
Synonyms: ethyl 3-(2-methoxypyridin-3-yl)prop-2-enoate
Molecular Weight (g/mol): 207.23
Molecular Formula: C11H13NO3
InChi Key: IUQBGDTUXITMKK-UHFFFAOYSA-N
SMILES: CCOC(=O)C=CC1=C(OC)N=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results