accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3COONHNOCH3CH3H3C(E/Z)

CAS RN 912760-85-1

IUPAC Name: ethyl 3-[4-(2,2-dimethylpropanamido)pyridin-3-yl]prop-2-enoate
Synonyms: ethyl 3-[4-(2,2-dimethylpropanamido)pyridin-3-yl]prop-2-enoate
Molecular Weight (g/mol): 276.34
Molecular Formula: C15H20N2O3
InChi Key: DVOJEHIQGVFQJY-UHFFFAOYSA-N
SMILES: CCOC(=O)C=CC1=C(NC(=O)C(C)(C)C)C=CN=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results