accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 912569-67-6

CH3NNONHH3C

CAS RN 912569-67-6

IUPAC Name: methyl({4-[(6-methylpyrazin-2-yl)oxy]phenyl}methyl)amine
Synonyms: methyl({4-[(6-methylpyrazin-2-yl)oxy]phenyl}methyl)amine
Molecular Weight (g/mol): 229.28
Molecular Formula: C13H15N3O
InChi Key: DAHTXAIQFDPRFK-UHFFFAOYSA-N
SMILES: CNCC1=CC=C(OC2=NC(C)=CN=C2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results