accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3NNSOOCl

CAS RN 912569-59-6

IUPAC Name: 3-(1-methyl-1H-pyrazol-3-yl)benzene-1-sulfonyl chloride
Synonyms: 3-(1-methylpyrazol-3-yl)benzenesulfonyl chloride
Molecular Weight (g/mol): 256.7
Molecular Formula: C10H9ClN2O2S
InChi Key: QGUULJKRHYTFTP-UHFFFAOYSA-N
SMILES: CN1C=CC(=N1)C1=CC(=CC=C1)S(Cl)(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results