CAS RN 912569-58-5
IUPAC Name:
ethyl 2-(4-methyl-1,4-diazepan-1-yl)benzoate
Synonyms:
ethyl 2-(4-methyl-1,4-diazepan-1-yl)benzoate
Molecular Weight (g/mol):
262.35
Molecular Formula:
C15H22N2O2
InChi Key:
XAVHDPRFJKPWNA-UHFFFAOYSA-N
SMILES:
CCOC(=O)C1=CC=CC=C1N1CCCN(C)CC1
1
–
1
of
1
results
1
Ethyl 2-(4-methylperhydro-1,4-diazepin-1-yl)benzoate, 97%, Thermo Scientific™
CAS: 912569-58-5 Molecular Formula: C15H22N2O2 Molecular Weight (g/mol): 262.353 MDL Number: MFCD09025904 InChI Key: XAVHDPRFJKPWNA-UHFFFAOYSA-N Synonym: ethyl 2-4-methylperhydro-1,4-diazepin-1-yl benzoate,ethyl 2-4-methyl-1,4-diazepan-1-yl benzoate,ethyl 2-4-methyl-1,4-diazaperhydroepinyl benzoate PubChem CID: 18525910 IUPAC Name: ethyl 2-(4-methyl-1,4-diazepan-1-yl)benzoate SMILES: CCOC(=O)C1=CC=CC=C1N2CCCN(CC2)C