accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 91215-97-3

H3CNCH3OH2N

CAS RN 91215-97-3

IUPAC Name: 1-{2-[2-(dimethylamino)ethoxy]phenyl}methanamine
Synonyms: 1-{2-[2-(dimethylamino)ethoxy]phenyl}methanamine
Molecular Weight (g/mol): 194.28
Molecular Formula: C11H18N2O
InChi Key: NOZRJUDRDVKQDW-UHFFFAOYSA-N
SMILES: CN(C)CCOC1=CC=CC=C1CN

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results