CAS RN 91183-98-1
CAS RN 91183-98-1
IUPAC Name:
{[5-(2,4-dioxo-1,2,3,4-tetrahydropyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methoxy}[({[3-acetamido-4,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}(hydroxy)phosphoryl)oxy]phosphinic acid disodium
Synonyms:
[5-(2,4-dioxo-3H-pyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methoxy({[3-acetamido-4,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy(hydroxy)phosphoryl}oxy)phosphinic acid disodium
Molecular Weight (g/mol):
653.33
Molecular Formula:
C17H27N3Na2O17P2
InChi Key:
QPWKUQXSMYTKRZ-UHFFFAOYNA-N
SMILES:
[Na].[Na].CC(=O)NC1C(O)C(O)C(CO)OC1OP(O)(=O)OP(O)(=O)OCC1OC(C(O)C1O)N1C=CC(=O)NC1=O
1
–
1
of
1
results
1
UDP-GlcNAc Disodium Salt, MedChemExpress
MedChemExpress UDP-GlcNAc Disodium Salt (UDP-α-D-N-Acetylglucosamine Disodium Salt) is a donor substrate of O-GlcNAc transferase (OGT).