accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

PNH2PNH2ClRuCl

CAS RN 911128-25-1

IUPAC Name: bis((3-aminopropyl)diphenylphosphane); dichlororuthenium
Synonyms: bis((3-aminopropyl)diphenylphosphane); dichlororuthenium
Molecular Weight (g/mol): 658.55
Molecular Formula: C30H36Cl2N2P2Ru
InChi Key: MLZFSEGMJUGESY-UHFFFAOYSA-L
SMILES: Cl[Ru]Cl.NCCCP(C1=CC=CC=C1)C1=CC=CC=C1.NCCCP(C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results