accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 910036-99-6

CH3HNNOO

CAS RN 910036-99-6

IUPAC Name: methyl({[6-(oxan-4-yloxy)pyridin-3-yl]methyl})amine
Synonyms: methyl({[6-(oxan-4-yloxy)pyridin-3-yl]methyl})amine
Molecular Weight (g/mol): 222.29
Molecular Formula: C12H18N2O2
InChi Key: CWRQCHDTRDIGCU-UHFFFAOYSA-N
SMILES: CNCC1=CN=C(OC2CCOCC2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results