accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

OONO

CAS RN 910036-95-2

IUPAC Name: 6-(oxan-4-yloxy)pyridine-3-carbaldehyde
Synonyms: 6-(oxan-4-yloxy)pyridine-3-carbaldehyde
Molecular Weight (g/mol): 207.23
Molecular Formula: C11H13NO3
InChi Key: ZQQOWSDTLFHTAC-UHFFFAOYSA-N
SMILES: O=CC1=CN=C(OC2CCOCC2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results