accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 906352-99-6

H3COOONNCH3

CAS RN 906352-99-6

IUPAC Name: ethyl 3-[(6-methylpyrazin-2-yl)oxy]benzoate
Synonyms: ethyl 3-[(6-methylpyrazin-2-yl)oxy]benzoate
Molecular Weight (g/mol): 258.28
Molecular Formula: C14H14N2O3
InChi Key: ALDBUDYDJSLBJO-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC(OC2=NC(C)=CN=C2)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results