accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 906352-80-5

OONCl

CAS RN 906352-80-5

IUPAC Name: 5-(chloromethyl)-2-(oxan-4-yloxy)pyridine
Synonyms: 5-(chloromethyl)-2-(oxan-4-yloxy)pyridine
Molecular Weight (g/mol): 227.69
Molecular Formula: C11H14ClNO2
InChi Key: XPOHCIVJYJZKHS-UHFFFAOYSA-N
SMILES: ClCC1=CN=C(OC2CCOCC2)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)