accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 906352-79-2

OONOH

CAS RN 906352-79-2

IUPAC Name: [6-(oxan-4-yloxy)pyridin-3-yl]methanol
Synonyms: [6-(oxan-4-yloxy)pyridin-3-yl]methanol
Molecular Weight (g/mol): 209.25
Molecular Formula: C11H15NO3
InChi Key: UTEOAQJLPJOMJS-UHFFFAOYSA-N
SMILES: OCC1=CN=C(OC2CCOCC2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results