accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

ClNS

CAS RN 906352-60-1

IUPAC Name: 2-[3-(chloromethyl)phenyl]-1,3-thiazole
Synonyms: 2-[3-(chloromethyl)phenyl]-1,3-thiazole
Molecular Weight (g/mol): 209.69
Molecular Formula: C10H8ClNS
InChi Key: SWICJACRAPFUKX-UHFFFAOYSA-N
SMILES: ClCC1=CC=CC(=C1)C1=NC=CS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results