accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 906352-58-7

NNSOOFFFFF

CAS RN 906352-58-7

IUPAC Name: 2,3,4,5,6-pentafluorophenyl 2-(pyridin-3-yl)-1,3-thiazole-4-carboxylate
Synonyms: 2,3,4,5,6-pentafluorophenyl 2-(pyridin-3-yl)-1,3-thiazole-4-carboxylate
Molecular Weight (g/mol): 372.27
Molecular Formula: C15H5F5N2O2S
InChi Key: LHRLGPDYSCUADI-UHFFFAOYSA-N
SMILES: FC1=C(F)C(F)=C(OC(=O)C2=CSC(=N2)C2=CN=CC=C2)C(F)=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results