accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 904816-32-6

NHHN

CAS RN 904816-32-6

IUPAC Name: 2-(naphthalen-2-yl)piperazine
Synonyms: 2-(naphthalen-2-yl)piperazine
Molecular Weight (g/mol): 212.3
Molecular Formula: C14H16N2
InChi Key: DNMNNHMPDTUAFG-UHFFFAOYNA-N
SMILES: C1CNC(CN1)C1=CC=C2C=CC=CC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)