CAS RN 90347-66-3
CAS RN 90347-66-3
IUPAC Name:
methyl 3-iodo-4-methylbenzoate
Synonyms:
methyl 3-iodo-4-methylbenzoate
Molecular Weight (g/mol):
276.07
Molecular Formula:
C9H9IO2
InChi Key:
NKMHAOTZPFVSPC-UHFFFAOYSA-N
SMILES:
COC(=O)C1=CC(I)=C(C)C=C1
1
–
1
of
1
results
1
Methyl 3-iodo-4-methylbenzoate, 98%
CAS: 90347-66-3 Molecular Formula: C9H9IO2 Molecular Weight (g/mol): 276.073 MDL Number: MFCD00230583 InChI Key: NKMHAOTZPFVSPC-UHFFFAOYSA-N Synonym: 3-iodo-4-methylbenzoic acid methyl ester,methyl 3-iodo-p-toluate,methyl-3-iodo-4-methylbenzoate,methyl 3-iodo-4-methyl-benzoate,2-iodo-4-methoxycarbonyl toluene,benzoic acid, 3-iodo-4-methyl-, methyl ester,methyl3-iodo-4-methylbenzoate,3-iodo-4-methylbenzoic acidmethyl ester,methyl 3-iodo-4-meth,ksc495m1r PubChem CID: 13614033 IUPAC Name: methyl 3-iodo-4-methylbenzoate SMILES: CC1=C(C=C(C=C1)C(=O)OC)I