accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 90325-47-6

H3C(S)OH(R)OHOH

CAS RN 90325-47-6

IUPAC Name: (2R,3S)-hexane-1,2,3-triol
Synonyms: (2R,3S)-hexane-1,2,3-triol
Molecular Weight (g/mol): 134.18
Molecular Formula: C6H14O3
InChi Key: XYXCXCJKZRDVPU-NTSWFWBYSA-N
SMILES: CCC[C@H](O)[C@H](O)CO
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)