accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3HNFOFF

CAS RN 902836-70-8

IUPAC Name: {[2-(2,4-difluorophenoxy)-5-fluorophenyl]methyl}(methyl)amine
Synonyms: {[2-(2,4-difluorophenoxy)-5-fluorophenyl]methyl}(methyl)amine
Molecular Weight (g/mol): 267.25
Molecular Formula: C14H12F3NO
InChi Key: MFAJRYWQXVCHHW-UHFFFAOYSA-N
SMILES: CNCC1=C(OC2=C(F)C=C(F)C=C2)C=CC(F)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results