accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 902836-38-8

HNNNCl

CAS RN 902836-38-8

IUPAC Name: 4-[4-(4-chlorophenyl)-1H-pyrazol-1-yl]piperidine
Synonyms: 4-[4-(4-chlorophenyl)pyrazol-1-yl]piperidine
Molecular Weight (g/mol): 261.75
Molecular Formula: C14H16ClN3
InChi Key: QPXZSTSEYCYKLI-UHFFFAOYSA-N
SMILES: ClC1=CC=C(C=C1)C1=CN(N=C1)C1CCNCC1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results