accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 902835-88-5

ONO

CAS RN 902835-88-5

IUPAC Name: 2-(cyclopropylmethoxy)pyridine-3-carbaldehyde
Synonyms: 2-(cyclopropylmethoxy)pyridine-3-carbaldehyde
Molecular Weight (g/mol): 177.2
Molecular Formula: C10H11NO2
InChi Key: AEOXNPSSZSDLDK-UHFFFAOYSA-N
SMILES: O=CC1=C(OCC2CC2)N=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results