accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 90134-10-4

H3CNCH3O

CAS RN 90134-10-4

IUPAC Name: 4-(dibutylamino)benzaldehyde
Synonyms: 4-(dibutylamino)benzaldehyde
Molecular Weight (g/mol): 233.36
Molecular Formula: C15H23NO
InChi Key: VNWPLOWYHIDMEB-UHFFFAOYSA-N
SMILES: CCCCN(CCCC)C1=CC=C(C=O)C=C1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)