CAS RN 9001-40-5
CAS RN 9001-40-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
2
Glucose-6-phosphate dehydrogenase, from Leuconostoc mesenteroides, ammonium sulfate suspension, Thermo Scientific™
CAS: 9001-40-5 MDL Number: MFCD00081656 Synonym: E.C.1.1.1.49