accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 90-90-4

IUPAC Name: (4-bromophenyl)(phenyl)methanone
Synonyms: (4-bromophenyl)(phenyl)methanone
Molecular Weight (g/mol): 261.12
Molecular Formula: C13H9BrO
InChi Key: KEOLYBMGRQYQTN-UHFFFAOYSA-N
SMILES: BrC1=CC=C(C=C1)C(=O)C1=CC=CC=C1
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results