accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3OOINOO

CAS RN 89976-27-2

IUPAC Name: methyl 4-iodo-3-nitrobenzoate
Synonyms: methyl 4-iodo-3-nitrobenzoate
Molecular Weight (g/mol): 307.04
Molecular Formula: C8H6INO4
InChi Key: SCMBIQRYVKITCY-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=C(I)C(=C1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results