accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3HNOO

CAS RN 898289-40-2

IUPAC Name: methyl({[4-(oxan-4-yloxy)phenyl]methyl})amine
Synonyms: methyl({[4-(oxan-4-yloxy)phenyl]methyl})amine
Molecular Weight (g/mol): 221.3
Molecular Formula: C13H19NO2
InChi Key: BIMJTFGNKMMZOM-UHFFFAOYSA-N
SMILES: CNCC1=CC=C(OC2CCOCC2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results