accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3H3CCH3PNNNNH3CH3CCH3(?)

CAS RN 894086-00-1

IUPAC Name: 5-(di-tert-butylphosphanyl)-1',3',5'-triphenyl-1'H-1,4'-bipyrazole
Synonyms: 5-(di-tert-butylphosphanyl)-1',3',5'-triphenyl-1,4'-bipyrazole
Molecular Weight (g/mol): 506.63
Molecular Formula: C32H35N4P
InChi Key: PTXJGGGNGMPMBG-UHFFFAOYSA-N
SMILES: CC(C)(C)P(C1=CC=NN1C1=C(N(N=C1C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1)C(C)(C)C
Molecular Weight (g/mol) 
  • (1)
  • (2)
Physical Form 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)