accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 89353-42-4

IUPAC Name: 3-(1H-pyrrol-1-yl)aniline
Synonyms: 3-(pyrrol-1-yl)aniline
Molecular Weight (g/mol): 158.2
Molecular Formula: C10H10N2
InChi Key: PJGDCPOPSNUYHC-UHFFFAOYSA-N
SMILES: NC1=CC=CC(=C1)N1C=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results