accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 89279-16-3

NH2OClCl

CAS RN 89279-16-3

IUPAC Name: 2-(2,4-dichlorophenoxy)aniline
Synonyms: 2-(2,4-dichlorophenoxy)aniline
Molecular Weight (g/mol): 254.11
Molecular Formula: C12H9Cl2NO
InChi Key: NXXSJWHUACPWNZ-UHFFFAOYSA-N
SMILES: NC1=CC=CC=C1OC1=CC=C(Cl)C=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results