accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

OHNN

CAS RN 892502-12-4

IUPAC Name: [3-(pyrimidin-2-yl)phenyl]methanol
Synonyms: [3-(pyrimidin-2-yl)phenyl]methanol
Molecular Weight (g/mol): 186.21
Molecular Formula: C11H10N2O
InChi Key: WMNXGJMCXOLBBV-UHFFFAOYSA-N
SMILES: OCC1=CC=CC(=C1)C1=NC=CC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results