accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3CNHNN

CAS RN 892502-08-8

IUPAC Name: methyl({4-[(1H-pyrazol-1-yl)methyl]phenyl}methyl)amine
Synonyms: methyl({[4-(pyrazol-1-ylmethyl)phenyl]methyl})amine
Molecular Weight (g/mol): 201.27
Molecular Formula: C12H15N3
InChi Key: BQLXMVISWVMAID-UHFFFAOYSA-N
SMILES: CNCC1=CC=C(CN2C=CC=N2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results