accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3HNNO

CAS RN 892502-04-4

IUPAC Name: {[5-(furan-2-yl)pyridin-2-yl]methyl}(methyl)amine
Synonyms: {[5-(furan-2-yl)pyridin-2-yl]methyl}(methyl)amine
Molecular Weight (g/mol): 188.23
Molecular Formula: C11H12N2O
InChi Key: DOKOUBYFAJTDEI-UHFFFAOYSA-N
SMILES: CNCC1=NC=C(C=C1)C1=CC=CO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results