accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 89-72-5

CH3CH3HO

CAS RN 89-72-5

IUPAC Name: 2-(butan-2-yl)phenol
Synonyms: 2-sec-butylphenol 2-sec.butylfenol
Molecular Weight (g/mol): 150.22
Molecular Formula: C10H14O
InChi Key: NGFPWHGISWUQOI-UHFFFAOYNA-N
SMILES: CCC(C)C1=CC=CC=C1O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results